accessibility menu, dialog, popup

UserName

CAS RN 351002-90-9

CHN

CAS RN 351002-90-9

IUPAC Name: 2-(4-ethynylphenyl)acetonitrile
Synonyms: 2-(4-ethynylphenyl)acetonitrile
Molecular Weight (g/mol): 141.17
Molecular Formula: C10H7N
InChi Key: CZUBOLAETAAHLA-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results