missing translation for 'onlineSavingsMsg'
Learn More

4-Ethynylphenylacetonitrile, 95%, Thermo Scientific™

Product Code. 15437628
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437628 25 g 25g
15427628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437628 Supplier Thermo Scientific Alfa Aesar Supplier No. H61122.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351002-90-9
Molecular Formula C10H7N
Molecular Weight (g/mol) 141.17
MDL Number MFCD05664204
InChI Key CZUBOLAETAAHLA-UHFFFAOYSA-N
Synonym 4-ethynylphenylacetonitrile, 2-4-ethynylphenyl acetonitrile, 4-ethynylphenyl acetonitrile, benzeneacetonitrile, 4-ethynyl, 4-ethynylbenzeneacetonitrile, ksc497s1l
PubChem CID 16069314
IUPAC Name 2-(4-ethynylphenyl)acetonitrile
SMILES C#CC1=CC=C(CC#N)C=C1

Specifications

Density 1.037 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 25 g
UN Number UN3276
Solubility Information Slightly soluble in water.
Formula Weight 141.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Ethynylphenylacetonitrile
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.