missing translation for 'onlineSavingsMsg'
Learn More

TTP 22, MedChemExpress

Product Code. 16432657
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16432657

Brand: Medchem Express HY15479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TTP 22 is a potent CK2 inhibitor, with an IC50 of 100 nM and a Ki of 40 nM.

TTP 22 is a potent CK2 inhibitor, with an IC50 of 100 nM and a Ki of 40 nM.

Specifications

Chemical Name or Material TTP 22
Physical Form Solid
CAS 329907-28-0
Color Gray
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H14N2O2S2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 51 mg/mL (154.35 mM)
SMILES O=C(O)CCSC1=C(C(C2=CC=C(C)C=C2)=CS3)C3=NC=N1
Molecular Weight (g/mol) 330.42
Formula Weight 330.42
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.