accessibility menu, dialog, popup

UserName

CAS RN 325-89-3

OHOH2NF

CAS RN 325-89-3

IUPAC Name: 3-amino-3-(4-fluorophenyl)propanoic acid
Synonyms: 3-amino-3-(4-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: CPGFMWPQXUXQRX-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)