accessibility menu, dialog, popup

UserName

CAS RN 32332-98-2

ONNH

CAS RN 32332-98-2

IUPAC Name: 5-(furan-2-yl)-1H-pyrazole
Synonyms: 3-(furan-2-yl)-2H-pyrazole
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: XHTWJTDNZKWQON-UHFFFAOYSA-N
SMILES: N1N=CC=C1C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results