accessibility menu, dialog, popup

UserName

CAS RN 32137-73-8

H3CSiCH3CH3NS

CAS RN 32137-73-8

IUPAC Name: 2-(trimethylsilyl)-1,3-benzothiazole
Synonyms: 2-(trimethylsilyl)-1,3-benzothiazole
Molecular Weight (g/mol): 207.37
Molecular Formula: C10H13NSSi
InChi Key: MNXBVXVIRAIAEG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=NC2=CC=CC=C2S1
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)