CAS RN 30754-23-5
IUPAC Name:
5,10,15,20,25,30,35-heptakis(iodomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Synonyms:
5,10,15,20,25,30,35-heptakis(iodomethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Weight (g/mol):
1904.27
Molecular Formula:
C42H63I7O28
InChi Key:
YODKQJPYQJRHEQ-UHFFFAOYNA-N
SMILES:
OC1C(O)C2OC3OC(CI)C(OC4OC(CI)C(OC5OC(CI)C(OC6OC(CI)C(OC7OC(CI)C(OC8OC(CI)C(OC1OC2CI)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
1
–
1
of
1
results
1
Heptakis-6-iodo-6-deoxy-β-cyclodextrin, Thermo Scientific™
CAS: 30754-23-5 Molecular Formula: C42H63I7O28 Molecular Weight (g/mol): 1904.28 MDL Number: MFCD00803290 InChI Key: YODKQJPYQJRHEQ-FOUAGVGXSA-N Synonym: heptakis-6-iodo-6-deoxy-beta-cyclodextrin PubChem CID: 11105283 SMILES: C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CI)CI)CI)CI)CI)CI)O)O)I