missing translation for 'onlineSavingsMsg'
Learn More

Heptakis-6-iodo-6-deoxy-β-cyclodextrin, Thermo Scientific™

Product Code. 10461491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10461491 100 mg Glass bottle 100mg
10115370 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10461491 Supplier Thermo Scientific Acros Supplier No. 299011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30754-23-5
Molecular Formula C42H63I7O28
Molecular Weight (g/mol) 1904.28
MDL Number MFCD00803290
InChI Key YODKQJPYQJRHEQ-FOUAGVGXSA-N
Synonym heptakis-6-iodo-6-deoxy-beta-cyclodextrin
PubChem CID 11105283
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CI)CI)CI)CI)CI)CI)O)O)I

Specifications

Melting Point 218°C to 222°C
Color White
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf,dmso,hot methylethyleneglycol
Formula Weight 1904.28
Physical Form Solid
Chemical Name or Material Heptakis-6-iodo-6-deoxy-β-cyclodextrin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.