CAS RN 3054-01-1
CAS RN 3054-01-1
IUPAC Name:
2-amino-3-(benzylsulfanyl)propanoic acid
Synonyms:
benzylcysteine
Molecular Weight (g/mol):
211.28
Molecular Formula:
C10H13NO2S
InChi Key:
GHBAYRBVXCRIHT-UHFFFAOYNA-N
SMILES:
NC(CSCC1=CC=CC=C1)C(O)=O
1
–
4
of
4
results
1
S-Benzyl-L-cysteine, 99%, Thermo Scientific Chemicals
CAS: 3054-01-1 MDL Number: MFCD00002613
2
S-Benzyl-L-cysteine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
S-Benzyl-L-cysteine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
S-Benzyl-L-cysteine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.