missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Benzyl-L-cysteine, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | S-Benzyl-L-cysteine |
| CAS | 3054-01-1 |
| Molecular Formula | C10H13NO2S |
| Quantity | 10 mg |
| Synonym | Alanine, 3-(benzylthio)-, L- (8CI), S-(Phenylmethyl)-L-cysteine, (2R)-2-Amino-3-(benzylsulfanyl)propanoic acid, (2R)-2-Amino-3-benzylsulfanylpropanoic acid, (R)-2-Amino-3-(benzylthio)propanoic acid, (R)-S-Benzylcysteine, (S)-S-Benzylcysteine, 3-Benzylthioalanine, L-S-Benzylcysteine, NSC 523123, S-Benzyl-(R)-cysteine, S-Benzyl-L-cysteine, S-Benzylcysteine |
| InChI Formula | InChI=1S/C10H13NO2S/c11-9(10(12)13)7-14-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)/t9-/m0/s1 |
| SMILES | N[C@@H](CSCc1ccccc1)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-benzylsulfanylpropanoic acid |
| Molecular Weight (g/mol) | 211.28 |
| Formula Weight | 211.0667 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction