CAS RN 288401-64-9
IUPAC Name:
6-(2,3-dihydro-1H-pyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
Synonyms:
6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
Molecular Weight (g/mol):
178.17
Molecular Formula:
C9H7FN2O
InChi Key:
DIVXDXZKHCYBPY-UHFFFAOYSA-N
SMILES:
FC1=CC(=C2NNC=C2)C(=O)C=C1
1
–
1
of
1
results
1
4-Fluoro-2-(1H-pyrazol-3-yl)phenol, 97%, Thermo Scientific™
CAS: 288401-64-9 Molecular Formula: C9H7FN2O Molecular Weight (g/mol): 178.17 InChI Key: DIVXDXZKHCYBPY-FPLPWBNLSA-N Synonym: 4-fluoro-2-1h-pyrazol-3-yl phenol,3-5-fluoro-2-hydroxyphenyl pyrazole,pyrazole, 31,6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one PubChem CID: 16217010 IUPAC Name: (6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one SMILES: C1=CC(=O)C(=C2C=CNN2)C=C1F