missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(1H-pyrazol-3-yl)phenol, 97%, Thermo Scientific™

Product Code. 11916221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916221 5 g 5g
11906221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916221 Supplier Thermo Scientific Acros Supplier No. 439060050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 288401-64-9
Molecular Formula C9H7FN2O
Molecular Weight (g/mol) 178.17
InChI Key DIVXDXZKHCYBPY-FPLPWBNLSA-N
Synonym 4-fluoro-2-1h-pyrazol-3-yl phenol, 3-5-fluoro-2-hydroxyphenyl pyrazole, pyrazole, 31, 6-3z-1,2-dihydropyrazol-3-ylidene-4-fluorocyclohexa-2,4-dien-1-one
PubChem CID 16217010
IUPAC Name (6Z)-6-(1,2-dihydropyrazol-3-ylidene)-4-fluorocyclohexa-2,4-dien-1-one
SMILES C1=CC(=O)C(=C2C=CNN2)C=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 132.9°C to 134.1°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 178.17
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-2-(1H-pyrazol-3-yl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.