accessibility menu, dialog, popup

UserName

CAS RN 278603-08-0

H3CNNCH3I

CAS RN 278603-08-0

IUPAC Name: 3-benzyl-1-hexadecyl-2-methyl-1H-imidazol-3-ium iodide
Synonyms: 1-benzyl-3-hexadecyl-2-methylimidazol-1-ium iodide
Molecular Weight (g/mol): 524.58
Molecular Formula: C27H45IN2
InChi Key: RVWOHCBHAGBLLT-UHFFFAOYSA-M
SMILES: [I-].CCCCCCCCCCCCCCCCN1C=C[N+](CC2=CC=CC=C2)=C1C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results