CAS RN 2758431-93-3
CAS RN 2758431-93-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Pomalidomide 5'-piperazine-4-methylpiperidine
Cereblon ligand with terminal piperidine for onward chemistry