accessibility menu, dialog, popup

UserName

CAS RN 27561-62-2

ONNH

CAS RN 27561-62-2

IUPAC Name: 1-cyclohexanecarbonylpiperazine
Synonyms: 1-cyclohexanecarbonylpiperazine
Molecular Weight (g/mol): 196.29
Molecular Formula: C11H20N2O
InChi Key: ZSZROXCAFYZNHE-UHFFFAOYSA-N
SMILES: O=C(C1CCCCC1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results