accessibility menu, dialog, popup

UserName

CAS RN 26964-24-9

CH3OOO

CAS RN 26964-24-9

IUPAC Name: 6-methoxy-2-phenyl-4H-chromen-4-one
Synonyms: 6-methoxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 252.27
Molecular Formula: C16H12O3
InChi Key: XZQLSABETMKIGG-UHFFFAOYSA-N
SMILES: COC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results