CAS RN 258843-62-8
CAS RN 258843-62-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Hoechst 33258 analog
MedChemExpress Hoechst 33258 analog are part of a family of blue fluorescent dyes used to stain DNA.