accessibility menu, dialog, popup

UserName

CH3CH3OOHHNSOOFFF

CAS RN 251097-65-1

IUPAC Name: 4-methyl-2-[3-(trifluoromethyl)benzenesulfonamido]pentanoic acid
Synonyms: 4-methyl-2-[3-(trifluoromethyl)benzenesulfonamido]pentanoic acid
Molecular Weight (g/mol): 339.33
Molecular Formula: C13H16F3NO4S
InChi Key: PGVOZRMIHHGLDI-UHFFFAOYNA-N
SMILES: CC(C)CC(NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)