missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-leucine, 96%, Thermo Scientific™

Product Code. 15418705
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418705 250 mg 250mg
15408705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418705 Supplier Thermo Scientific Alfa Aesar Supplier No. H33922.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251097-65-1
Molecular Formula C13H16F3NO4S
Molecular Weight (g/mol) 339.33
MDL Number MFCD00173033
InChI Key PGVOZRMIHHGLDI-UHFFFAOYNA-N
Synonym 4-methyl-2-3-trifluoromethyl phenylsulfonamido pentanoic acid, 4-methyl-2-3-trifluoromethyl benzenesulfonamido pentanoic acid, 3-trifluoromethyl phenyl sulfonyl leucine, leucine,n-3-trifluoromethyl phenyl sulfonyl, 4-methyl-2-3-trifluoromethyl phenyl sulfonyl amino pentanoic acid, 4-methyl-2-3-trifluoromethyl benzenesulphonylamino pentanoic acid, 4-methyl-2-3-trifluoromethyl phenyl-sulfonyl amino pentanoic acid, 4-methyl-2-3-trifluoromethyl phenyl sulfonylamino pentanoic acid
PubChem CID 2775543
IUPAC Name 4-methyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]pentanoic acid
SMILES CC(C)CC(NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F)C(O)=O

Specifications

Melting Point 123°C to 124°C
Quantity 250 mg
Formula Weight 339.34
Percent Purity 96%
Chemical Name or Material N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.