CAS RN 2435-36-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl cyclopentane-1,3-dicarboxylate, TRC
CAS: 2435-36-1 Molecular Formula: C9H14O4 Molecular Weight (g/mol): 186.21 IUPAC Name: dimethyl cyclopentane-1,3-dicarboxylate SMILES: COC(=O)C1CCC(C1)C(=O)OC