missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cyclopentane-1,3-dicarboxylate, TRC

Product Code. 30383695
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383695 100 mg 100mg
30383491 50 mg -
30375981 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383695 Supplier Toronto Research Chemicals Supplier No. TRCD482063100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2435-36-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
IUPAC Name dimethyl cyclopentane-1,3-dicarboxylate
SMILES COC(=O)C1CCC(C1)C(=O)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H14O4/c1-12-8(10)6-3-4-7(5-6)9(11)13-2/h6-7H,3-5H2,1-2H3
Formula Weight 186.0892
Chemical Name or Material Dimethyl cyclopentane-1,3-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.