CAS RN 232271-19-1
CAS RN 232271-19-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TR-14035, MedChemExpress
MedChemExpress TR-14035 is a orally active dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune diseases.