missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR-14035, MedChemExpress
Description
TR-14035 is a orally active dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune diseases.
Specifications
Specifications
| Chemical Name or Material | TR-14035 |
| CAS | 232271-19-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H21Cl2NO5 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 41 mg/mL (86.44 mM) |
| SMILES | COC1=C(C2=CC=C(C[C@@H](C(O)=O)NC(C3=C(Cl)C=CC=C3Cl)=O)C=C2)C(OC)=CC=C1 |
| Molecular Weight (g/mol) | 474.33 |
| Formula Weight | 474.33 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?