accessibility menu, dialog, popup

UserName

BrBrPP

CAS RN 22884-31-7

IUPAC Name: triphenyl[5-(triphenylphosphaniumyl)pentyl]phosphanium dibromide
Synonyms: triphenyl[5-(triphenylphosphaniumyl)pentyl]phosphanium dibromide
Molecular Weight (g/mol): 754.53
Molecular Formula: C41H40Br2P2
InChi Key: XIZMFZWZYVQIOX-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results