missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylenebis(triphenylphosphonium bromide), 98+%, Thermo Scientific™

Product Code. 11334414
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334414 25 g 25g
11324414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334414 Supplier Thermo Scientific Alfa Aesar Supplier No. L00997.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pentamethylenebis(triphenylphosphonium bromide), is used as an intermediate in Pharmaceutical research.

Solubility
It is soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. Moisture sensitive. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-31-7
Molecular Formula C41H40Br2P2
Molecular Weight (g/mol) 754.53
MDL Number MFCD00051887
InChI Key XIZMFZWZYVQIOX-UHFFFAOYSA-L
Synonym pentane-1,5-diylbis triphenylphosphonium bromide, pentamethylenebis triphenylphosphonium bromide, triphenyl 5-triphenylphosphaniumylpentyl phosphanium dibromide, triphenyl 5-triphenylphosphaniumyl pentyl phosphanium dibromide, 1,5-pentanediylbis triphenylphosphonium dibromide, 1,1'-1,5-pentanediyl bis 1,1,1-triphenyl-, bromide 1:2
PubChem CID 3084382
IUPAC Name triphenyl(5-triphenylphosphaniumylpentyl)phosphanium;dibromide
SMILES [Br-].[Br-].C(CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 253°C to 260°C
Quantity 25 g
Beilstein 3875042
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 754.54
Percent Purity ≥98%
Chemical Name or Material Pentamethylenebis(triphenylphosphonium bromide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.