CAS RN 2244386-63-6
CAS RN 2244386-63-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Pravastatin sodium salt hydrate
CAS: 2244386-63-6 Molecular Formula: C23H35NaO7 Molecular Weight (g/mol): 446.52 MDL Number: MFCD00887601 InChI Key: VWBQYTRBTXKKOG-IYNICTALSA-M IUPAC Name: sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate SMILES: [Na+].[H][C@]12[C@H](C[C@H](O)C=C1C=C[C@H](C)[C@@H]2CC[C@@H](O)C[C@@H](O)CC([O-])=O)OC(=O)[C@@H](C)CC