accessibility menu, dialog, popup

UserName

CAS RN 22424-58-4

CH3ONOO

CAS RN 22424-58-4

IUPAC Name: 4-(benzyloxy)-2-methyl-1-nitrobenzene
Synonyms: 4-(benzyloxy)-2-methyl-1-nitrobenzene
Molecular Weight (g/mol): 243.26
Molecular Formula: C14H13NO3
InChi Key: PBAXHOUGHZKSRP-UHFFFAOYSA-N
SMILES: CC1=CC(OCC2=CC=CC=C2)=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results