accessibility menu, dialog, popup

UserName

CH3NHNH2NOO

CAS RN 22280-62-2

IUPAC Name: 6-amino-2-methyl-3-nitropyridin-1-ium
Synonyms: 6-amino-2-methyl-3-nitropyridin-1-ium
Molecular Weight (g/mol): 154.15
Molecular Formula: C6H8N3O2
InChi Key: BGMZTBKXOFFTBJ-UHFFFAOYSA-O
SMILES: CC1=[NH+]C(N)=CC=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)