missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-3-nitro-2-picoline, 98%, Thermo Scientific Chemicals

Product Code. 10590352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10590352 1 g Glass bottle 1g
10390194 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10590352 Supplier Thermo Scientific Acros Supplier No. 360770010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22280-62-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153
MDL Number MFCD00053582
InChI Key BGMZTBKXOFFTBJ-UHFFFAOYSA-N
Synonym 2-amino-6-methyl-5-nitropyridine, 6-amino-3-nitro-2-picoline, 2-amino-5-nitro-6-methylpyridine, 6-amino-2-methyl-3-nitropyridine, 2-amino-5-nitro-6-methyl pyridine, 2-amino-5-nitro-6-picoline, 6-amino-3-nitro-2-methylpyridine, 6-methyl-5-nitro-2-pyridinamine, 6-methyl-5-nitro-pyridin-2-ylamine, 6-methyl-5-nitro-2-pyridylamine
PubChem CID 247976
IUPAC Name 6-methyl-5-nitropyridin-2-amine
SMILES CC1=C(C=CC(=N1)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 185°C to 189°C
Color Beige to Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Amino-3-nitro-2-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-3-nitro-2-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.