CAS RN 2224-02-4
CAS RN 2224-02-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Asparagusic acid, MedChemExpress
MedChemExpress Asparagusic acid is a sulfur-containing flavor component produced by asparagus plants, with anti-parasitic effect. Asparagusic acid is a plant growth inhibitor.