missing translation for 'onlineSavingsMsg'
Learn More

Asparagusic acid, MedChemExpress

Product Code. 16450558
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450558

Brand: Medchem Express HY50730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asparagusic acid is a sulfur-containing flavor component produced by asparagus plants, with anti-parasitic effect. Asparagusic acid is a plant growth inhibitor.

Asparagusic acid is a sulfur-containing flavor component produced by asparagus plants, with anti-parasitic effect. Asparagusic acid is a plant growth inhibitor.

Specifications

Chemical Name or Material Asparagusic acid
Physical Form Solid
CAS 2224-02-4
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C4H6O2S2
Solubility Information DMSO : ≥ 46 mg/mL (306.22 mM)
SMILES O=C(O)C1CSSC1
Molecular Weight (g/mol) 150.22
Formula Weight 150.22
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.