accessibility menu, dialog, popup

UserName

CAS RN 22148-86-3

H3COH

CAS RN 22148-86-3

IUPAC Name: 4-phenylbutan-2-ol
Synonyms: 2-hydroxy-4-phenylbutane
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: GDWRKZLROIFUML-UHFFFAOYNA-N
SMILES: CC(O)CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results