CAS RN 216766-12-0
CAS RN 216766-12-0
IUPAC Name:
6-(trifluoromethyl)pyridin-3-ol
Synonyms:
6-(trifluoromethyl)pyridin-3-ol
Molecular Weight (g/mol):
163.1
Molecular Formula:
C6H4F3NO
InChi Key:
VFOBDHYPESAMAF-UHFFFAOYSA-N
SMILES:
OC1=CN=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
CAS: 216766-12-0 Molecular Formula: C6H4F3NO Molecular Weight (g/mol): 163.10 MDL Number: MFCD06801325 InChI Key: VFOBDHYPESAMAF-UHFFFAOYSA-N Synonym: 6-trifluoromethyl pyridin-3-ol,5-hydroxy-2-trifluoromethyl pyridine,6-trifluoromethyl-3-pyridinol,2-trifluoromethyl pyridin-5-ol,5-hydroxy-2-trifluoromethylpyridine,3-pyridinol, 6-trifluoromethyl,6-trifluoromethyl-pyridin-3-ol,pubchem21818,6-trifluoromethyl 3-pyridinol PubChem CID: 2783388 IUPAC Name: 6-(trifluoromethyl)pyridin-3-ol SMILES: OC1=CN=C(C=C1)C(F)(F)F