accessibility menu, dialog, popup

UserName

CAS RN 215178-39-5

NHOOO

CAS RN 215178-39-5

IUPAC Name: (9H-fluoren-9-yl)methyl 3-hydroxypyrrolidine-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 3-hydroxypyrrolidine-1-carboxylate
Molecular Weight (g/mol): 309.37
Molecular Formula: C19H19NO3
InChi Key: KPDNZJQVYQCDJW-UHFFFAOYNA-N
SMILES: OC1CCN(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)