CAS RN 211516-63-1
CAS RN 211516-63-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oleanolic acid derivative 2, MedChemExpress
MedChemExpress Oleanolic acid derivative 2 is an oleanolic acid derivative, which is a novel triterpenoid-steroid hybrid molecule.