missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic acid derivative 2, MedChemExpress

Product Code. 16409927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 16409927

Brand: Medchem Express HY18003

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Oleanolic acid derivative 2 is an oleanolic acid derivative, which is a novel triterpenoid-steroid hybrid molecule.

Oleanolic acid derivative 2 is an oleanolic acid derivative, which is a novel triterpenoid-steroid hybrid molecule.

Specifications

Chemical Name or Material Oleanolic acid derivative 2
CAS 211516-63-1
Color White
Purity Grade Notes Research
Molecular Formula C28H42O3
Quantity 5 mg
SMILES C[C@@]12CC[C@](C(CC3)=O)([H])[C@@]3(C)[C@@]1([H])CC=C4[C@]5([H])CC(C)(C)CC[C@@](C(OC)=O)5CC[C@@]24C
Molecular Weight (g/mol) 426.63
Formula Weight 426.63
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.