CAS RN 210095-58-2
CAS RN 210095-58-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-[(S)-(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine L-Tartrate, TRC
CAS: 210095-58-2 Molecular Formula: C17 H19 Cl N2 O . C4 H6 O6 Molecular Weight (g/mol): 452.89 Synonym: 2-[(S)-(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine (2R,3R)-2,3-Dihydroxybutanedioate IUPAC Name: 2-[(S)-(4-chlorophenyl)-piperidin-4-yloxymethyl]pyridine;(2R,3R)-2,3-dihydroxybutanedioic acid SMILES: O[C@H]([C@@H](O)C(=O)O)C(=O)O.Clc1ccc(cc1)[C@H](OC2CCNCC2)c3ccccn3