missing translation for 'onlineSavingsMsg'
Learn More

2-[(S)-(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine L-Tartrate, TRC

Product Code. 30368450
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368450 100 g 100g
30358963 5 g 5g
30353283 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368450 Supplier Toronto Research Chemicals Supplier No. TRCC380375100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210095-58-2
Molecular Formula C17 H19 Cl N2 O . C4 H6 O6
Molecular Weight (g/mol) 452.89
Synonym 2-[(S)-(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine (2R,3R)-2,3-Dihydroxybutanedioate
IUPAC Name 2-[(S)-(4-chlorophenyl)-piperidin-4-yloxymethyl]pyridine;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES O[C@H]([C@@H](O)C(=O)O)C(=O)O.Clc1ccc(cc1)[C@H](OC2CCNCC2)c3ccccn3

Specifications

Quantity 100 g
InChI Formula InChI=1S/C17H19ClN2O.C4H6O6/c18-14-6-4-13(5-7-14)17(16-3-1-2-10-20-16)21-15-8-11-19-12-9-15;5-1(3(7)8)2(6)4(9)10/h1-7,10,15,17,19H,8-9,11-12H2;1-2,5-6H,(H,7,8)(H,9,10)/t17-;1-,2-/m01/s1
Formula Weight 452.135
Chemical Name or Material 2-[(S)-(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine L-Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.