CAS RN 2084868-04-0
CAS RN 2084868-04-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ Capzimin dimer
Selective proteasome inhibitor
2
Capzimin, MedChemExpress
MedChemExpress Capzimin is a potent and moderately specific proteasome isopeptidase Rpn11 inhibitor.