CAS RN 2070014-83-2
CAS RN 2070014-83-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chitinase-IN-2 hydrochloride, MedChemExpress
MedChemExpress Chitinase-IN-2 hydrochloride is a insect chitinase and N- acetyl hexosaminidase inhibitor and pesticide; 50 uM/20uM compound concentration's inhibitory percentage are 98%/92% for chitinase/N- acetyl-hexosaminidase respectively.