accessibility menu, dialog, popup

UserName

CAS RN 203314-27-6

CH3FOHO

CAS RN 203314-27-6

IUPAC Name: 2-(2-fluoro-5-methylphenyl)acetic acid
Synonyms: (2-fluoro-5-methylphenyl)acetic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: CQFSSMCJXPLZAT-UHFFFAOYSA-N
SMILES: CC1=CC(CC(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results