accessibility menu, dialog, popup

UserName

CAS RN 199590-75-5

CH3ClSOONH2OCH3

CAS RN 199590-75-5

IUPAC Name: 5-chloro-2-methoxy-4-methylbenzene-1-sulfonamide
Synonyms: 5-chloro-2-methoxy-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 235.68
Molecular Formula: C8H10ClNO3S
InChi Key: VQHIRMCXZNAVRG-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Cl)C(C)=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results