CAS RN 1992047-65-0
CAS RN 1992047-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PPTN hydrochloride
High affinity and selective P2Y14 antagonist