missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PPTN hydrochloride

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
PPTN hydrochloride is a high affinity and selective P2Y14 antagonist (KB = 434 pM), which exhibits >10,000-fold selectivity for P2Y14 over other P2Y receptors. PPTN inhibits UDP-glucose and MRS 2690 -induced porcine pancreatic artery contraction ex vivo, and also blocks UDP-glucose-induced chemotaxis of HL-60 leukemia cells in vitro.
Specifications
Specifications
| Chemical Name or Material | 4-[4-(4-Piperidinyl)phenyl]-7-[4-(trifluoromethyl)phenyl]-2-naphthalenecarboxylic acid hydrochloride |
| CAS | 1992047-65-0 |
| Quantity | 10 mg |
| Target | P2Y Receptor Antagonists |
| Molecular Formula | C29H24F3NO2.HCl |
| Formula Weight | 511.96 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction