missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Altrose, 99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| CAS | 1990-29-0 |
| Melting Point | 106°C to 108°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (HPLC) |
| Packaging | Glass bottle |
| Molecular Formula | C6H12O6 |
| MDL Number | MFCD00075620 |
| Quantity | 100 mg |
| Merck Index | 15, 317 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction