accessibility menu, dialog, popup

UserName

H3CNNOOOCH3O

CAS RN 191846-76-1

IUPAC Name: 5-methoxy-1-methyl-4-nitro-1H-indole-3-carbaldehyde
Synonyms: 5-methoxy-1-methyl-4-nitroindole-3-carbaldehyde
Molecular Weight (g/mol): 234.21
Molecular Formula: C11H10N2O4
InChi Key: YDMDOIHSAYQATQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2N(C)C=C(C=O)C2=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results