missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde, 90+%, Thermo Scientific™

Product Code. 11324156
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324156 250 mg 250mg
11304156 1 g 1g
11314156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324156 Supplier Thermo Scientific Alfa Aesar Supplier No. L19270.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191846-76-1
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD03093098
InChI Key YDMDOIHSAYQATQ-UHFFFAOYSA-N
Synonym 5-methoxy-1-methyl-4-nitroindole-3-carboxaldehyde, 5-methoxy-1-methyl-4-nitro-1h-indole-3-carbaldehyde, 1-methyl-5-methoxy-4-nitro-indole-3-carboxaldehyde, acmc-20am2n
PubChem CID 10847280
IUPAC Name 5-methoxy-1-methyl-4-nitroindole-3-carbaldehyde
SMILES CN1C=C(C2=C1C=CC(=C2[N+](=O)[O-])OC)C=O

Specifications

Melting Point ∽190°C (decomposition)
Quantity 250 mg
Beilstein 7766233
Sensitivity Air Sensitive
Formula Weight 234.21
Percent Purity ≥90%
Chemical Name or Material 5-Methoxy-1-methyl-4-nitroindole-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.