accessibility menu, dialog, popup

UserName

CAS RN 191338-86-0

CH3H3CONHOHOOHNH3COHONHOHOOHNOCH3

CAS RN 191338-86-0

IUPAC Name: 4-({1-[(1-carboxy-3-oxopropan-2-yl)carbamoyl]-2-hydroxypropyl}carbamoyl)-4-(2-acetamido-3-methylpentanamido)butanoic acid
Synonyms: 4-({1-[(1-carboxy-3-oxopropan-2-yl)carbamoyl]-2-hydroxypropyl}carbamoyl)-4-(2-acetamido-3-methylpentanamido)butanoic acid
Molecular Weight (g/mol): 502.52
Molecular Formula: C21H34N4O10
InChi Key: AXTKTZHLZLOIIO-UHFFFAOYNA-N
SMILES: CCC(C)C(NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(C(C)O)C(=O)NC(CC(O)=O)C=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)