accessibility menu, dialog, popup

UserName

CAS RN 190788-60-4

BOOH3CCH3H3CH3COH3C

CAS RN 190788-60-4

IUPAC Name: 2-(2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 234.1
Molecular Formula: C13H19BO3
InChi Key: ASDFSWMHZSWXPO-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results