missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMD 3465 hexahydrobromide, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AMD 3465 hexahydrobromide (GENZ-644494 hexahydrobromide) is a potent antagonist of CXCR4, inhibits binding of 12G5 mAb and CXCL12AF647 to CXCR4, with IC50s of 0.75 nM and 18 nM in SupT1 cells
Specifications
Specifications
| Chemical Name or Material | AMD 3465 hexahydrobromide |
| Physical Form | Solid |
| CAS | 185991-07-5 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H44Br6N6 |
| For Use With (Application) | COVID-19-anti-virus |
| Synonym | GENZ-644494 hexahydrobromide |
| Solubility Information | DMSO : 50 mg/mL (55.80 mM; Need ultrasonic) ∣H2O : ≥ 38 mg/mL (42.41 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction